• USA Home
  • C1650 - Chitinase from Serratia marcescens

EMAIL THIS PAGE TO A FRIEND
C1650 Sigma

Chitinase from Serratia marcescens

lyophilized powder, ≥10 units/g solid

Synonym: Chitodextrinase, Poly(1,4-β-[2-acetamido-2-deoxy-D-glucoside]) glycanohydrolase

Purchase

Description

Other Notes

View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer

Unit Definition

One unit will liberate 1.0 mg of N-acetyl-D-glucosamine from chitin per hour at pH 6.0 at 25 °C in a 2 hour assay.
One new 1 hour unit = approx. 50 old 48 hour units.

Physical form

Lyophilized powder containing phosphate buffer salts

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
FM6205000
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C8241 Chitinase from Trichoderma viride, lyophilized powder, ≥600 units/g solid

substrate

Product #

Description

Add to Cart

C3020 Chitin azure, chitinase substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?