• USA Home
  • C1658 - α-Casein Fragment 90-95

EMAIL THIS PAGE TO A FRIEND
C1658 Sigma

α-Casein Fragment 90-95

≥97% (HPLC)

Synonym: Arg-Tyr-Leu-Gly-Tyr-Leu

  • CAS Number 83471-50-5

  • Empirical Formula (Hill Notation) C38H57N9O9

  • Molecular Weight 783.91

  •  MDL number MFCD00076691

  •  PubChem Substance ID 24892430

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Enzyme Fragments, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   KWLNZVXBGCEDOO-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?