• USA Home
  • C1662 - (−)-1,6-Bis(2-chlorophenyl)-1,6-diphenyl-2,4-hexadiyne-1,6-diol

EMAIL THIS PAGE TO A FRIEND
C1662 Sigma

(−)-1,6-Bis(2-chlorophenyl)-1,6-diphenyl-2,4-hexadiyne-1,6-diol

  • CAS Number 96158-84-8

  • Empirical Formula (Hill Notation) C30H20Cl2O2

  • Molecular Weight 483.38

  •  MDL number MFCD00135912

  •  PubChem Substance ID 24892431

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   CJETXAUUVYDHTN-KYJUHHDHSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?