• USA Home
  • C1748 - Anti-CLK1 (86-100) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C1748 Sigma

Anti-CLK1 (86-100) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-CDC-like kinase 1, Anti-CLK/STY, Anti-CLK, Anti-STY

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CI-CN, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~57 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CLK1(1195)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 86-100 of human CLK1

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-CLK1 (86-100) antibody produced in rabbit is suitable for western blotting at a dilution of 1:500-1:2000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

CDC-like kinase 1 (CLK1) is an enzyme encoded by the CLK1 gene in humans. It belongs to the CDC2-like (or LAMMER) family of dual specificity protein kinases. It is also referred as CLK and STY. Clk/Sty belongs to Clk/Sty family of dual-specificity kinases. It is autophosphorylated using both direct kinase assays and SR protein-dependent splicing assays on both Ser/Thr and Thr residues. Clk/Sty recognize different SR proteins, which is influenced by the extent and nature of autophosphorylation. It is found in the nucleus of several different cell types. CLK expression had distinct effects on HIV-1, CLK1 enhances Gag production while CLK2 inhibits the virus.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

SAB1300108 Anti-CLK1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?