• USA Home
  • C1785 - Cibacron Blue 3G-A–Dextran

EMAIL THIS PAGE TO A FRIEND
C1785 Sigma

Cibacron Blue 3G-A–Dextran

Synonym: Formerly listed as Reactive Blue 2-Dextran

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
extent of labeling   50-100 μmol per g
storage temp.   2-8°C

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?