EMAIL THIS PAGE TO A FRIEND
C1857 Sigma

Citric acid

meets USP testing specifications, anhydrous

Purchase

Properties

grade   anhydrous
InChI Key   KRKNYBCHXYNGOX-UHFFFAOYSA-N
assay   99.5-100.5% anhydrous basis
expl. lim.   8 %, 65 °F
pKa    (1) 3.13, (2) 4.76, (3) 6.4
mp   153-159 °C(lit.)
suitability   meets USP testing specifications

Description

Application

In addition to its function as an iron chelator, citrate can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications.

Price and Availability


Interactive Buffer Table

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
GE7350000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?