• USA Home
  • C1883 - Ammonium citrate dibasic

EMAIL THIS PAGE TO A FRIEND
C1883 Sigma

Ammonium citrate dibasic

~98% (capillary GC)

Synonym: Ammonium hydrogencitrate, Citric acid ammonium salt, Diammonium hydrogen citrate

  • CAS Number 3012-65-5

  • Linear Formula HOC(CO2H)(CH2CO2NH4)2

  • Molecular Weight 226.18

  •  Beilstein Registry Number 4925760

  •  EC Number 221-146-3

  •  MDL number MFCD00013068

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffers A to Z, Chelators
vapor density   1.8 (vs air)
InChI Key   YXVFQADLFFNVDS-UHFFFAOYSA-N
assay   ~98% (capillary GC)
pH   5.2 (20 °C, 50 g/L)
Featured Industry   Diagnostic Assay Manufacturing

Price and Availability


Interactive Buffer Table
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
GE7545000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

247561 Ammonium citrate dibasic, ACS reagent, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?