• USA Home
  • C188 - 6-Hydroxychlorzoxazone

EMAIL THIS PAGE TO A FRIEND
C188 Sigma

6-Hydroxychlorzoxazone

solid

Synonym: 5-Chloro-6-hydroxy-2(3H)-benzoxazolone, 5-Chloro-6-hydroxybenzoxazone

  • CAS Number 1750-45-4

  • Empirical Formula (Hill Notation) C7H4ClNO3

  • Molecular Weight 185.56

  •  MDL number MFCD00143502

Purchase

Properties

InChI Key   AGLXDWOTVQZHIQ-UHFFFAOYSA-N
form   solid
solubility   H2O: insoluble
  ethanol: moderately soluble

Description

Application

CYP2E1 & 1A2 metabolite of chlorzoxazone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DM5260000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

UC148 6-Hydroxychlorzoxazone, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?