• USA Home
  • C1978 - Anti-Calcium Channel (β3 Subunit) (Cacnb3l) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C1978 Sigma

Anti-Calcium Channel (β3 Subunit) (Cacnb3l) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Calcium Channels, Antibodies to Ion Channels,
species reactivity   rat
application(s)   immunohistochemistry: suitable
  western blot (chemiluminescent): 1:200-1:300
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... Cacnb3(25297)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 463-477 of the β3 subunit of rat brain voltage-gated calcium channel (VGCC, Caβ3) with additional N-terminal cysteine, conjugated to KLH. This epitope is identical in all known alternative splicing isoforms of Caβ3 in mouse, rat, human, and Xenopus.

Physical form

Lyophilized at approximately 0.3 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?