EMAIL THIS PAGE TO A FRIEND
C1983 Sigma

Cloflubicyne

≥95% (NMR), solid

  • Empirical Formula (Hill Notation) C11H6Cl2F6N2

  • Molecular Weight 351.08

  •  MDL number MFCD00236471

  •  PubChem Substance ID 24892456

Purchase

Description

General description

Potent non-competitive GABA antagonist; convulsant; laboratory insecticide.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?