EMAIL THIS PAGE TO A FRIEND
C2005 Sigma

Coprostane

Synonym: 5β-Cholestane

  • CAS Number 481-20-9

  • Empirical Formula (Hill Notation) C27H48

  • Molecular Weight 372.67

  •  MDL number MFCD00067141

  •  PubChem Substance ID 24892458

Purchase

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?