• USA Home
  • C211 - Corticotropin Releasing Factor human, rat

EMAIL THIS PAGE TO A FRIEND
C211 Sigma

Corticotropin Releasing Factor human, rat

Synonym: CRF, CRH

  • CAS Number 86784-80-7

  • Empirical Formula (Hill Notation) C208H344N60O63S2

  • Molecular Weight 4757.45

  •  MDL number MFCD00213806

Purchase

Properties

storage temp.   −20°C

Description

Amino Acid Sequence

Ser-Glu-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-Leu-Met-Glu-Ile-Ile-NH2

Biochem/physiol Actions

CRF is a hypothalamic peptide that releases ACTH and endorphin from the anterior pituitary. It is also a neurotransmitter in the central nervous system, and is involved in autonomic and endocrine responses to stress.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GM7925000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C3042 Corticotropin Releasing Factor human, rat, ≥95% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?