EMAIL THIS PAGE TO A FRIEND
C212 Sigma

N-Acetyl-L-cysteine

Synonym: LNAC, NAC

  • CAS Number 616-91-1

  • Linear Formula HSCH2CH(NHCOCH3)CO2H

  • Molecular Weight 163.19

  •  Beilstein Registry Number 1724426

  •  EC Number 210-498-3

  •  MDL number MFCD00004880

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   PWKSKIMOESPYIA-BYPYZUCNSA-N
mp   106-108 °C(lit.)
storage temp.   −20°C

Description

Biochem/physiol Actions

Antioxidant and mucolytic agent. Increases cellular pools of free radical scavengers. Reported to prevent apoptosis in neuronal cells but induce apoptosis in smooth muscle cells. Inhibits HIV replication. May serve as a substrate for microsomal glutathione transferase.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
HA1660000
Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

A7250 N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?