• USA Home
  • C226 - Monoclonal Anti-Calcitonin Gene-Related Peptide antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C226 Sigma

Monoclonal Anti-Calcitonin Gene-Related Peptide antibody produced in mouse

clone C4/19, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-CGRP

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CA1-CAL, Calcitonin and Related Products, Cell Biology,
biological source   mouse
antibody form   purified immunoglobulin
clone   C4/19, monoclonal
form   buffered aqueous solution
species reactivity   rat, human
application(s)   dot blot: 1:10,000 using rat CGRP-BSA
  immunohistochemistry: 1:8,000 using 4% paraformaldehyde perfusin/rat brain
  radioimmunoassay: 1:5,000 using 125I-CGRP (rat)
isotype   IgG1
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Immunogen

rat α-CGRP conjugated to BSA

Preparation Note

The antibody is purified using protein A agarose, 0.2 μm-filtered and supplied at approximately 1 mg/ml.

Specificity

The antibody reacts by ELISA with rat α- and β-CGRP and human α- and β-CGRP. By RIA the antibody has a KD = 1.9 nM. The antibody may be used to localize and detect calcitonin gene-related peptide (CGRP) -containing axon terminals in the cortex, caudate putamen and cell bodies, axons and terminals of CGRP-containing neurons in the CNS and periphery.

Physical form

Solution in 0.01 M phosphate buffered saline, containing 0.1% sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?