EMAIL THIS PAGE TO A FRIEND
C2418 Sigma
Reswelling cassette
Purchase
Description
Application
Specifically designed for rehydrating Immobiline DryPlates to proper thickness. During reswelling, additives can be incorporated into the gel.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?