EMAIL THIS PAGE TO A FRIEND
C2524 Sigma

Cyclo(Phe-Ser)

≥97% (TLC)

  • CAS Number 35591-00-5

  • Empirical Formula (Hill Notation) C12H14N2O3

  • Molecular Weight 234.25

  •  MDL number MFCD00056023

  •  PubChem Substance ID 24892499

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   WIJKWEYCUNYTGY-UWVGGRQHSA-N
assay   ≥97% (TLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Intermediate in the biosynthesis of gliotoxin by Trichoderma viride.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?