• USA Home
  • C2629 - Acetylcholinesterase from Electrophorus electricus (electric eel)

EMAIL THIS PAGE TO A FRIEND
C2629 Sigma

Acetylcholinesterase from Electrophorus electricus (electric eel)

Type III, aqueous solution, ≥1,000 units/mg protein

Synonym: AChE, Acetylcholine acetylhydrolase, Cholinesterase, Acetyl

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience,
type   Type III
form   aqueous solution
impurities   ~5 mg/mg protein (NH4)2SO4
color   clear to slightly hazy light yellow
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

The activity obtained using acetylcholine as substrate is 30-100 times that obtained with butyrylcholine, using acetylcholinesterase from electric eel.

Biochem/physiol Actions

Major degradative enzyme for acetylcholine in vivo. Converts acetylcholine + H2O to choline + acetic acid.

Caution

Excellent stability while frozen; avoid repeated freeze-thaw cycles.

Unit Definition

One unit will hydrolyze 1.0 μmole of acetylcholine to choline and acetate per min at pH 8.0 at 37 °C.

General description

Molecular Weight: 280 kDa
Isoelectric Point: 5.5
Extinction Coefficient: E1% = 18.0 (280 nm)

Acetylcholinesterase from Electrophorus electricus is a tetramer composed of 4 equal subunits of 70 kDa each. Each subunit contains one active site. The enzyme is a glycoprotein containing hexosamines.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

C2888 Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein

inhibitor

Product #

Description

Add to Cart

D7910 3,4-Dichloroisocoumarin, serine protease inhibitor
H5777 (±)-Huperzine A, synthetic, ≥98% (TLC)
A9013 1,5-Bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide
A3773 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?