• USA Home
  • C263 - Cytokine-Induced Neutrophil Chemoattractant-2 from rat

EMAIL THIS PAGE TO A FRIEND
C263 Sigma

Cytokine-Induced Neutrophil Chemoattractant-2 from rat

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested

Synonym: CINC-2α, CINC-2

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemokines, Cytokine-Induced Neutrophil Chemoattractant, Cytokines and Growth Factors,
recombinant   expressed in E. coli
assay   ≥97% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
suitability   cell culture tested
storage temp.   −20°C
Gene Information   rat ... Cxcl3(171551)

Description

Physical form

Lyophilized from 30% acetonitrile and 0.1% trifluoroacetic acid containing 1.25 mg bovine serum albumin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?