• USA Home
  • C2733 - 5(6)-Carboxyfluorescein, succinimidyl

EMAIL THIS PAGE TO A FRIEND
C2733 Sigma

5(6)-Carboxyfluorescein, succinimidyl

Synonym: FLUOS

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?