• USA Home
  • C2835 - CHO Serum-free Medium without L-Glutamine

EMAIL THIS PAGE TO A FRIEND
C2835 Sigma

CHO Serum-free Medium without L-Glutamine

Purchase

Properties

Quality Level   GMP
storage temp.   2-8°C

Description

Formula variant

Without fetuin and L-glutamine.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C5467 EX-CELL® ACF CHO Medium, Animal-component free, with HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?