• USA Home
  • C2858 - 5(6)-Carboxytetramethylrhodamine

EMAIL THIS PAGE TO A FRIEND
C2858 Sigma

5(6)-Carboxytetramethylrhodamine

Purchase

Properties

InChI Key   LBUFYZITRTUEFI-UHFFFAOYSA-N
mp   ≥300 °C(lit.)

Description

Application

Reacts with primary and secondary amine groups on proteins, nucleic acids and other biomolecules.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?