EMAIL THIS PAGE TO A FRIEND
C2980 Sigma

Coelenterazine f

solid

Synonym: CLZN-f

Purchase

Description

Biochem/physiol Actions

Almost 20 times higher luminescence intensity and an emission maximum which is 8 nm longer than native coelenterazine (Prod. No. C 2230).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?