• USA Home
  • C2983 - 5(6)-Carboxy-X-rhodamine N-succinimidyl ester

EMAIL THIS PAGE TO A FRIEND
C2983 Sigma

5(6)-Carboxy-X-rhodamine N-succinimidyl ester

  • CAS Number 114616-32-9

  • Empirical Formula (Hill Notation) C37H33N3O7

  • Molecular Weight 631.67

Purchase

Properties

InChI Key   YEWCZAUCUAYJIO-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?