• USA Home
  • C3006 - Cytochrome c from bovine heart

EMAIL THIS PAGE TO A FRIEND
C3006 Sigma

Cytochrome c from bovine heart

practical grade, ≥60% based on Mol. Wt. 12,327 basis

Purchase

Properties

Related Categories Cell Biology, Cytochrome c, Cytochromes, Electron Transport and Cellular Respiration, Metabolic Pathways,
grade   practical grade
assay   ≥60% based on Mol. Wt. 12,327 basis
mol wt   mol wt 12327 Da
storage temp.   −20°C
Gene Information   cow ... CYC1(512500)

Description

Other Notes

View more information on cytochrome c and electron transport at www.sigma-aldrich.com/enzymeexplorer.

Preparation Note

Prepared without using TCA.

Quality

Contains variable amounts of reduced cytochrome c.

Biochem/physiol Actions

The ready interconversion of cytochrome c between ferrous and ferric states makes it an efficient biological electron carrier. It plays a vital role in cellular oxidations in both plants and animals. Generally regarded as a universal link in the respiratory chain, it forms the essential electron-bridge between the respirable substrates and oxygen.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HA5365000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO PolyMA-SCX (Ion Exchange)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO PolyMA-SCX (Ion Exchange)
Keywords: Chromatography, High performance liquid chromatography, Separation

HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO Wide Pore C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO Wide Pore C18
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Proteins on Discovery® BIO PolyMA-SCX (Analyte Set #2)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Proteins on Discovery® BIO PolyMA-SCX (Analyte Set #2)
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

C3131 Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis

related product

Product #

Description

Add to Cart

C4186 Cytochrome c partially acetylated from equine heart, lyophilized powder
C2506 Cytochrome c from equine heart, ≥95% (SDS-PAGE)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?