• USA Home
  • C3101 - Anti-Caspase 8 (SK-16) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C3101 Sigma

Anti-Caspase 8 (SK-16) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Caspases,
species reactivity   human
application(s)   microarray: suitable
  western blot: 1:1,000 using human Burkitt lymphoma Raji cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CASP8(841)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 209-224 of the human caspase 8a/MACHα1 precursor, conjugated to KLH. The sequence contains the N-terminal region of the enzyme p18 subunit and is identical in other human caspase 8 isoforms (caspase 8b/MACHα2, MACHβ3, MACHβ4) and highly conserved (>85%) in mouse caspase 8.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Specificity

By immunoblotting, the antibody reacts with caspase 8a (55 kDa) and caspase 8b (54 kDa) and weakly cross-reacts with MACHβ3/MACHβ4 isoforms (30-32 kDa).

Price and Availability


Detect Your Unseen Protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?