• USA Home
  • C3166 - Lactocerebrosides from bovine brain

EMAIL THIS PAGE TO A FRIEND
C3166 Sigma

Lactocerebrosides from bovine brain

≥93%

Synonym: CDH, Ceramide β-lactoside, Ceramide dihexoside, Ceramide-β-D-lactoside

Purchase

Description

Packaging

1, 5 mg in glass bottle

Price and Availability


Amplified Detection

The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FK1000000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Glycosphingolipids

Glycosphingolipids (sometimes called glycolipids) have been found in lower and higher eukaryotic sources. They are composed of a glycan structure attached to a lipid tail that contains the sphingolip...
Glycobiology Analysis Manual, 2nd Edition
Keywords: Acetylations, Acylations, Cancer, Degradations, Gene expression, Growth factors, High performance liquid chromatography, Hydroxylations, Sulfations, Transduction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?