• USA Home
  • C3380 - Corynanthine hydrochloride

EMAIL THIS PAGE TO A FRIEND
C3380 Sigma

Corynanthine hydrochloride

Synonym: 17α-Hydroxyyohimban-16β-carboxylic acid methyl ester

Purchase

Properties

InChI Key   PIPZGJSEDRMUAW-VYDVLXQVSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ZG1025000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?