• USA Home
  • C3381 - Cytochrome c Reductase from porcine heart

EMAIL THIS PAGE TO A FRIEND
C3381 Sigma

Cytochrome c Reductase from porcine heart

Type I, lyophilized powder, ≥1.0 units/mg protein

Synonym: NADH dehydrogenase, NADH: (acceptor) oxidoreductase

Purchase

Description

Packaging

25, 100, 500 mg in poly bottle

Unit Definition

One unit will reduce 1.0 μmole of oxidized cytochrome c per min at pH 8.5 at 25 °C.

Physical form

Crude, lyophilized powder containing potassium phosphate, pH approx. 7.0

Biochem/physiol Actions

Cytochrome c reductase is a flavoprotein that completes the oxidation-reduction chain between hexosemonophosphate and cytochrome c. The molecular weight of cytochrome c reductase is found to be approximately 78,000 Da. The prosthetic group of cytochrome c reductase is alloxazine mononucleotide. The enzyme is very unstable with respect to low pH and to denaturation by heat.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?