• USA Home
  • C3392 - Monoclonal Anti-Caspase 4 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C3392 Sigma

Monoclonal Anti-Caspase 4 antibody produced in mouse

~2 mg/mL, clone CAS4, purified immunoglobulin, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Caspases,
species reactivity   monkey, human
application(s)   indirect ELISA: suitable
  microarray: suitable
  western blot: 1-2 μg/mL using a whole cell extract of monkey kidney cell line COS-7
clone   CAS4, monoclonal
concentration   ~2 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 43 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CASP4(837)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 119-133 of human caspase 4 protein conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?