• USA Home
  • C3540 - Polaroid® MP4+ sliding camera head
EMAIL THIS PAGE TO A FRIEND
C3540 Sigma
Polaroid® MP4+ sliding camera head
Purchase
Properties
availability   not available in EU
Description
Other Notes
Movable carriage that holds ground glass (G 7395) and reflex viewer/hood (R 6007). Allows focusing without removal of film holder from camera.
Legal Information
Polaroid is a registered trademark of Polaroid Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?