• USA Home
  • C3636 - Choloylglycine Hydrolase from Clostridium perfringens (C. welchii)

EMAIL THIS PAGE TO A FRIEND
C3636 Sigma

Choloylglycine Hydrolase from Clostridium perfringens (C. welchii)

crude acetone powder, 10-20 units/mg solid

Synonym: 3α,7α,12α-Trihydroxy-5β-cholan-24-oylglycine amidohydrolase

Purchase

Properties

Related Categories Cell Biology, General Metabolic Enzymes, General Metabolic Enzymes A-H, Metabolomics
form   crude acetone powder
storage temp.   −20°C

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of glycocholic acid to glycine and cholic acid in 5 min at pH 5.6 at 37°C.

Physical form

Crude acetone powder from which activity can easily be extracted

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C4018 Choloylglycine Hydrolase from Clostridium perfringens (C. welchii), lyophilized powder, ≥100 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?