• USA Home
  • C3724 - Hanna portable pH/conductivity/TDS/temperature meter
EMAIL THIS PAGE TO A FRIEND
C3724 Sigma
Hanna portable pH/conductivity/TDS/temperature meter
Hanna HI 991301 (high range)
Purchase
Properties
Related Categories Conductivity Meters, Hand Held, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories,
mfr. no.   Hanna HI 991301 (high range)
measuring range   0-10.00 ppt TDS, accuracy: 2%, resolution: 0.01 ppt
  0.0-60.0 °C temperature, accuracy: 0.5 °C, resolution: 0.1 °C
  0.00-14.00 pH, accuracy: 0.01, resolution: 0.01
  0.00-19.99 mS/cm conductivity, accuracy: 2%, resolution: 0.01 mS/cm
battery power   4 × 1.5 V AAA, 500 hr life
Description
General description
• Waterproof
• Single combination electrode
• Select °C or °F, Beta (ATC for conductivity) adjustable from 1.8 to 2.4%, ppm conversion factor from 0.45 to 0.74
• Simple two-button function control
• Illuminated % battery power display when unit is turned ON
• Complete with meter, probe, and rugged case
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?