• USA Home
  • C3762 - 1-Chloro-2,4-dinitrobenzene

EMAIL THIS PAGE TO A FRIEND
C3762 Sigma

1-Chloro-2,4-dinitrobenzene

~95%

Synonym: 2,4-Dinitrochlorobenzene, CDNB, DNCB

Purchase

Properties

InChI Key   VYZAHLCBVHPDDF-UHFFFAOYSA-N
assay   ~95%
expl. lim.   22 %
bp   315 °C(lit.)
mp   48-50 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 3441 6.1 / PGII
WGK Germany 
2
RTECS 
CZ0525000
Flash Point(F) 
381.2 °F
Flash Point(C) 
194 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

138630 1-Chloro-2,4-dinitrobenzene, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?