EMAIL THIS PAGE TO A FRIEND
C3782 Sigma

Cellulose acetate

Synonym: Acetylcellulose

Purchase

Properties

extent of labeling   ~40% acetyl
refractive index   n20/D 1.475(lit.)
density   1.3 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
nwg
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

180955 Cellulose acetate, average Mn ~30,000 by GPC

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?