• USA Home
  • C3856 - Greiner CapMat for sealing multiwell plates

EMAIL THIS PAGE TO A FRIEND
C3856 Sigma

Greiner CapMat for sealing multiwell plates

384 well CapMat, for sealing 384 well plates, square nap, translucent, piercebable

Synonym: Greiner CapMats for sealing multiwell plates, multiwell plate cap mats

Purchase

Properties

Related Categories 384 Well Plates, Cell Culture, Drug Discovery, Greiner, Greiner 384-Well Multiwell Plates,
material   silicone
sterility   non-sterile
packaging   pack of 10
  case of 50
mfr. no.   Greiner 384070
size   384 wells
color   translucent
matrix active group   Non-treated surface treatment
suitability   suitable for (for sealing 384 well plates)

Description

Chem/Phys Resistance

Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?