EMAIL THIS PAGE TO A FRIEND
C3926 Sigma
Casting clamps
For dual-gel casting assembly
Purchase
Description
General description
Specifically designed for gel casting. Wide jaws provide uniform pressure on glass plate-gasket assembly. Single- and dual-gel casting clamps accommodate different thicknesses and are not interchangeable.
Set of three (3). One set (three clamps) is sufficient for 10 cm wide mini-gels; two sets (six clamps) are necessary for standard 16.5 cm wide gels.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?