• USA Home
  • C3928 - Cryopreservation supplies
EMAIL THIS PAGE TO A FRIEND
C3928 Sigma
Cryopreservation supplies
Cup
Purchase
Properties
feature   capacity 7 straws
Description
Application
For holding cryopreservation straws in liquid nitrogen.
Compatibility
Rack (R 5011) is designed to suspend cup in liquid nitrogen.
General description
This product is a holding cup with a capacity to hold 7 cryopreservation straws.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?