• USA Home
  • C3960 - Anti-Ciliary Neurotrophic Factor antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
C3960 Sigma

Anti-Ciliary Neurotrophic Factor antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Description

Immunogen

recombinant human CNTF expressed in Escherichia coli.

General description

Ciliary neurotrophic factor is a cytokine of the IL-6 family and is crucial survival factor for ciliary ganglion neurons. The signalling initiated by CNTF is mediated by heteromeric receptor complex composed of CNTF receptor α, leukemia inhibitory factor receptor β and glycoprotein gp130. The binding of CNTF to the receptor complex results in the activation of pathways involving Jak and STAT family of DNA-binding transcription factors. The effective therapeutic use of CTNF has been studied in retinitis pigmentosa, Parkinson’s disease, Huntington’s disease and other neurodegenerative disorders
Anti-Human Ciliary Neurotrophic Factor (CNTF) antibody specifically recognizes human CNTF.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Specificity

The antibody neutralizes the biological activity of recombinant human CNTF.

Preparation Note

Purified using protein G.

Application

Anti-Human Ciliary Neurotrophic Factor (CNTF) antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. For ELISA a working concentration of 0.5-1.0 μg/ml may be used. The antibody is suitable for neutralization reactions.

Price and Availability


Detect Your Unseen Protein

All labs need water

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?