• USA Home
  • C4041 - 8-(3-Carboxypropyl)-1,3-dimethylxanthine

EMAIL THIS PAGE TO A FRIEND
C4041 Sigma

8-(3-Carboxypropyl)-1,3-dimethylxanthine

Synonym: Theophylline-8-butanoic acid

  • CAS Number 5438-71-1

  • Empirical Formula (Hill Notation) C11H14N4O4

  • Molecular Weight 266.25

  •  MDL number MFCD00055595

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   NGBYKZGOXFIONW-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XH5060000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?