• USA Home
  • C4085 - Anti-Ciliary Neurotrophic Factor antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
C4085 Sigma

Anti-Ciliary Neurotrophic Factor antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Purchase

Description

Immunogen

recombinant rat CNTF expressed in Escherichia coli.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Specificity

The antibody neutralizes the biological activity of recombinant rat CNTF.

Preparation Note

Purified using protein G.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?