• USA Home
  • C4126 - Z-Ala-Ser methyl ester

EMAIL THIS PAGE TO A FRIEND
C4126 Sigma

Z-Ala-Ser methyl ester

  • CAS Number 19542-34-8

  • Empirical Formula (Hill Notation) C15H20N2O6

  • Molecular Weight 324.33

  •  MDL number MFCD00056131

  •  PubChem Substance ID 24892664

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   IDVXGMLTOPUJTE-JQWIXIFHSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?