• USA Home
  • C4140 - Citrate Synthase from pigeon breast muscle

EMAIL THIS PAGE TO A FRIEND
C4140 Sigma

Citrate Synthase from pigeon breast muscle

ammonium sulfate suspension, 80-150 units/mg protein (modified Warburg-Christian)

Synonym: Citrate condensing enzyme, Citrate oxaloacetate lyase (CoA-acetylating)

Purchase

Properties

Related Categories 4.1.x.x C-C bonds, 4.x.x.x Lyases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   ammonium sulfate suspension
foreign activity   isocitrate dehydrogenase and aconitase ≤0.01%
  malic dehydrogenase ≤0.1%
storage temp.   2-8°C

Description

Unit Definition

One unit will form 1.0 μmole of citrate from oxalacetate and acetyl CoA per min at pH 8.0 at 37 °C.

Physical form

Crystalline suspension in 2.2 M (NH4)2SO4 solution, pH 7.0, containing 6 mM phosphate and 0.5 mM citrate

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Articles

Ammonium Sulfate Suspensions - Technical Note

Note that for an ammonium sulfate suspension, most of the enzyme will be in solid form. It is likely that only negligible amounts of enzyme will be in the ammonium sulfate solution.
Keywords: Enzyme activity

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Related Products

comparable product

Product #

Description

Add to Cart

C3260 Citrate Synthase from porcine heart, ammonium sulfate suspension, ≥100 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?