• USA Home
  • C4141 - Cyanomethyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside

EMAIL THIS PAGE TO A FRIEND
C4141 Sigma

Cyanomethyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside

  • CAS Number 61145-33-3

  • Empirical Formula (Hill Notation) C16H21NO9S

  • Molecular Weight 403.40

  •  MDL number MFCD00057536

  •  PubChem Substance ID 24892667

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Monosaccharide More...
InChI Key   ZNQCXCWNORCJGV-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?