EMAIL THIS PAGE TO A FRIEND
C4218 Sigma

Cycloheximide

plant cell culture tested, ~98% (TLC)

Synonym: 3-(2-[3,5-Dimethyl-2-oxocyclohexyl]-2-hydroxyethyl)glutarimide

Purchase

Properties

assay   ~98% (TLC)
suitability   plant cell culture tested
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
MA4375000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?