• USA Home
  • C4224 - HL-60 Cell Extract, Induced

EMAIL THIS PAGE TO A FRIEND
C4224 Sigma

HL-60 Cell Extract, Induced

solution in loading buffer

Synonym: HL-60 Cell Extract

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Immunohistology and Blotting Controls, Signal Transduction Research Tools
form   solution in loading buffer
suitability   suitable for positive control in immunoblotting techniques; useful as a control for PARP (Recommended use: 20 μl (about 75,000 cells) per gel lane.)

Description

Application

Control for Immunoblotting

General description

Whole cell extract of human HL-60 leukemia cells induced to undergo apoptosis using the chemotherapeutic agent etoposide.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?