EMAIL THIS PAGE TO A FRIEND
C4262 Sigma

Z-Gly-Tyr

~95%

  • CAS Number 7801-35-6

  • Empirical Formula (Hill Notation) C19H20N2O6

  • Molecular Weight 372.37

  •  MDL number MFCD00055742

  •  PubChem Substance ID 24892680

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   YYPNBRMRXNXSDE-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?