• USA Home
  • C4288 - βH-Crystallin from bovine eye lens

EMAIL THIS PAGE TO A FRIEND
C4288 Sigma

βH-Crystallin from bovine eye lens

Purchase

Properties

impurities   Salt, essentially free
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

C5163 βL-Crystallin from bovine eye lens

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?