EMAIL THIS PAGE TO A FRIEND
C4415 Sigma

trans-Cinnamic acid

plant cell culture tested

Synonym: β-Phenylacrylic acid, 3-Phenylpropenoic acid

Purchase

Properties

suitability   plant cell culture tested

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
GD7850000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?