EMAIL THIS PAGE TO A FRIEND
C4433 Sigma

Cys-Val-2-Nal-Met

≥90% (HPLC)

Purchase

Description

Amino Acid Sequence

Cys-Val-Ala(2-naphthyl)-Met

Biochem/physiol Actions

p21ras farnesyltransferase inhibitor. While many tetrapeptide "inhibitors" of p21ras FTase are known, most act instead as competitive substrates. This compound is not farnesylated and is thus a true inhibitor.

Price and Availability


All labs need water

Biomedical Applications

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?