• USA Home
  • C4481 - Anti-Caspase 4 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C4481 Sigma

Anti-Caspase 4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Caspases,
species reactivity   human
application(s)   immunoprecipitation: 5-10 μg using 300 μg lysate of human ECV304 endothelial cells.
  microarray: suitable
  western blot (chemiluminescent): 1:250 using whole cell extract of human ECV304 endotheloid cells
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 43-47 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CASP4(837)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to human procaspase 4α isoform sequence, (amino acids 81-98) with N-terminal lysine added, conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Application

Anti-Caspase 4 antibody produced in rabbit is suitable for:
• immunoprecipitation at a concentration of 5-10μg using 300μg lysate of human ECV304 endothelial cells.
• microarray
• western blotting (chemiluminescent)at a dilution of 1:250 using whole cell extract of human ECV304 endotheloid cells

Biochem/physiol Actions

Caspase-4 is a 377 amino acids protein "upstream" caspase having a long N-terminal prodomain. Caspase-4 is capable of cleaving itself and the p30 caspase-1 precursor, and of inducing apoptosis in transfected insect or COS cells.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?